Imprimer
Oral gavage needles, disposable
Fournisseur: CADENCE SCIENCE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9921 9918 9915 9920 9930 9931 9923 9924
POPR9921EA 2030 EUR
POPR9921 POPR9918 POPR9915 POPR9920 POPR9930 POPR9931 POPR9923 POPR9924
Oral gavage needles, disposable
Aiguilles pour seringues
Disposable sterile feeding needles are for critical applications.
  • All needles feature a plastic luer hub and a soft silicone tip
  • Malleable to any angle
  • Non-toxic and non-pyrogenic
  • Available with malleable stainless steel or flexible plastic (PTFE) tubing