Imprimer
Bag closures, WeLoc®
Fournisseur: WELOC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S1207001 S1206004 S1207004 S1211001 S1215001 S1222001 S1232001 S1206001_U S1207005 S1205004
312-0106EA 64.6 EUR
312-0106 312-0105 312-0107 312-0108 312-0109 312-0110 312-0111 WELOS1206001_U WELOS1207005 WELOS1205004
Bag closures, WeLoc®
Clamps General Purpose Clamps
These sealing clips are perfect for use in the demanding pharmaceutical and laboratory environment because they are made from high quality Polyamide 6.6 material from DuPont®. The clips help to maintain hygiene standards, securing opened bags of raw ingredients and materials, such as powders and fluids.
  • Re-closing of partly used bags of ingredients will reduce the risk of cross-contamination
  • Reduce the risk of leakage or accidents
  • Made from strong and durable material
  • Manufactured in Sweden under strict hygiene conditions
  • Clips can be sterilised via autoclave, EtO or gamma irradiation before being re-used