Imprimer
Immobilised avidin for biotin purification
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-594 786-593
786-594EA 1730 EUR
786-594 786-593
Immobilised avidin for biotin purification
Protein Purification Protein Purification Resins
This avidin resin is designed for the single step small and large scale affinity purification of proteins and antibodies with a biotin tag. The resin can also be used for immunoprecipitations using biotin labelled antibodies.