Imprimer
Cleaning solutions/spray for electrophoresis gel plates, GELPLATE-clean™
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-140 786-140RF 786-140RF-II 786-140RF-I
786-140EA 126 EUR
786-140 786-140RF 786-140RF-II 786-140RF-I
Cleaning solutions/spray for electrophoresis gel plates, GELPLATE-clean™
Produits nettoyants Décontaminants
GELPLATE-clean™ removes proteins, nucleic acids, fats, lipids, radioisotopes and other contaminants from electrophoresis gel plates. It is suitable for sequencing and protein electrophoresis gel plates.
  • Prevent poor electrophoretic band migration
  • Prevent band distortions
  • Prevent poor image development
Informations de livraison: Supplied as 2 solutions in 2× 250 ml spray bottles. Refill solutions are available and are offered separately.