Imprimer
Macroplate, 6-well
Fournisseur: Greiner bio-one
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
391-0263EA 202 EUR
391-0263
Macroplate, 6-well
Boîtes de culture Boîtes de Petri
Macroplate with Ø 35 mm per well, is a practical and space-saving alternative to six standard Petri dishes. Numeric coding allows easier identification of wells.