Imprimer
Wafer tweezers
Fournisseur: OUTILS RUBIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
43LB-8PEEK 43LB-4PEEK 43LB-4UPEEK 43LB-8UPEEK
OUTI43LB-8PEEKEA 50.1 EUR
OUTI43LB-8PEEK OUTI43LB-4PEEK OUTI43LB-4UPEEK OUTI43LB-8UPEEK
Wafer tweezers
Pinces
PEEK. These cleanroom compatible tweezers are specially designed to handle different fragile and delicate materials of gallium or silicone wafers.
  • Tips made of soft conductive fibre
  • Stopping pins turned up on lateral edges and graduated lower paddles prevent the wafer from slipping
  • ESD safe (105 Ω×cm)
  • Heat resistance up to 250 °C (short exposure 300 °C)
  • Not compatible with nitric and sulphuric acids