Imprimer
Multiple tissue blot, single species
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TB38 TB39
TB38EA 2090 EUR
TB38 TB39
Multiple tissue blot, single species
Réactifs d'électrophorèse Western Blotting Reagents
These multiple tissue, single species blots permit researchers to visualise the tissue distribution of their protein in a particular species. These have been successfully used for the analysis of a wide variety of proteins. Each blot has multiple tissue types from either human, mouse or rat species.