Imprimer
Biotin reagents, OneQuant™ HOOK™
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-083 786-084 786-085 786-698 786-699 786-700
786-083EA 433 EUR
786-083 786-084 786-085 786-698 786-699 786-700
Biotin reagents, OneQuant™ HOOK™
Systèmes de liaison et de marquage des protéines
The OneQuant™ format prevents loss of reagent due to repeated weighing as each vial contains only 1 - 2 mg HOOK™ Biotin Reagent.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13e8dfed1bf64442a7dceafe29e33a8d5d0865b1&vid=c3e1d5b8facee25cbe05c176c2d98ff8ce179b84&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df83296ec1e7f955f878dcb6206fda945643b803&vid=dc7eee65ee2cea60a57858f6ad79f40fcf6f0d16&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d540f70135ec6fb5ea5eaf26687f26494e02f880&vid=993d813daacc154489875b5faa1b9d377deb3caa&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9969b7e13a3833de7b2cac3797f32c573f6e0508&vid=4888f7f4492c11671b510e04ea2d76d6d5cef611&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d6aef171e1842d10a195e754a9342cf1cde383a&vid=63fbc1af319e5f9a094438285c08ce441ee1efc8&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59e20fd06123275c55f6d813d152a88f565191b7&vid=72ebb93b13ad8fafb69c35fde2ac8d65e6ed8ebb&prid=web&clid=SAPDAM