Imprimer
Chromatography tubing, stainless
Fournisseur: VICI JOUR RESEARCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JR-TSS.520-M3 JR-TSS.510-M3 JR-TSS140-M10
JOURJR-TSS.520-M3EA 239 EUR
JOURJR-TSS.520-M3 JOURJR-TSS.510-M3 JOURJR-TSS140-M10
Chromatography tubing, stainless
Tuyaux Tubulures à chromatographie
SS316, premium grade, seamless.
  • Smooth internal surface
  • Soft annealed OD for easy ferrule connections
A variety of sizes for HPLC and GC applications. Tubings can withstand a pressure rating of min 400 bar (5800 psi) and 365 bar (5300 psi)