Imprimer
Safety bunsen burner
Fournisseur: schuett-biotec GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3355202 3354202 3356202
SCUT3355202EA 1140 EUR
SCUT3355202 SCUT3354202 SCUT3356202
Safety bunsen burner
Appareils chauffants Brûleurs à gaz
Safety bunsen burner made of polished stainless steel and safety glass. Ideal for use in microbiological safety cabinets or in the chemical laboratory.
  • Multilingual menu navigation in most international languages
  • Easy-to-read highlighted colour display
  • Gas and air adjustment with “Click-Stop”-feature
  • Automated safety shutdown if not in use
  • Overheating protection/burner control by sensors activation 1x/2x
Schuett phoenix II eco: without display/sensor, with foot-switch
Schuett phoenix II standard: colour display, language selection, sensor and foot- switch
Schuett phoenix II accu: colour display, language selection, sensor and built-in high-power battery
W×D×H: 125×160×85 mm
Weight: 1000 g