Imprimer
Turbidity meters, MicroTol
Fournisseur: HF SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20053 20054 20055 20056 20063 20064
708-0022EA 0 EUR
708-0022 708-0023 708-0024 708-0025 708-0026 708-0027
Turbidity meters, MicroTol
Turbidimètres
MicroTol turbidity meter provides fast and easy calibration.
  • Verification in seconds while a complete primary calibration can be completed in less than five minutes
  • Low maintenance fail safe design
  • Simple modular design easy to use and service
  • Bubble rejection system
  • Eliminates bubbles without delaying the response time
  • Low volume sample chamber
  • Two user selectable high/low/system alarms
Measurement principle: Nephelometry (90°)
Standard outputs: 4 to 20 mA galvanic isolated or RS485 (selectable)
Certifications: White light version compliant to U.S. EPA 180.1; infrared version compliant to ISO 7027; CE approved, ETL listed to UL 61010-1 3rd Edition: 2012, ETL certified to CSA 22.2 No.61010-1-12: 2012.
Informations de commande: Ultrasonic autoclean system, and a data acquisition software system that allows logging and data storage for multiple turbidimeters.