Imprimer
Snap-off replacement blades for cutters
Fournisseur: WEDO WERNER DORSCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
789 7818
231-2902EA 14.6 EUR
231-2902 231-2906
Snap-off replacement blades for cutters
Rasoirs et lames
These snap-off replacement cutter blades are made of SK5 carbon steel and are suitable for almost all commercial cutter models.
  • High quality
  • High edge retention and sharpness
Cdt: Supplied in a plastic box with 10 blades.