Imprimer
Stands for 6 L Bio-bin® Boxes
Fournisseur: ECONIX LTD
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BEXBBZS6.0 BEXBBUS6.0
119-0230EA 429 EUR
119-0230 119-0229
Stands for 6 L Bio-bin® Boxes
Boxes Waste Disposal Boxes
These stands for 6 litre Bio-bin® waste disposal boxes are made from stainless steel.
Z stand: Stand holds the 6 L Bio-bin® upright, either for benchtop or floor use and allows for easy removal of used bins. Stand is supplied with 4 non slip feet for base of stand to prevent sliding.
U stand: The U stand holds the 6 L Bio-bin® at a 30º angle, making it perfect for use in a fume cupboard. Stand is supplied with 4 non slip feet for base of stand to prevent sliding.