Imprimer
Detergent removal systems, OrgoSol DetergentOUT™
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-127 786-128
786-127EA 433 EUR
786-127 786-128
Detergent removal systems, OrgoSol DetergentOUT™
Protéines extraction Réactifs pour l'extraction des protéines
OrgoSol DetergentOUT™ is suitable for removal of detergents from protein solutions, including hydrophobic protein solutions. Compatible with all detergent types, its performance is not dependent on the concentration of detergents in the solution. It is highly flexible and can be used to process small and large sample volumes.
OrgoSol DetergentOUT™ first concentrates the protein solution through precipitation and then the detergent is extracted and removed with the supplied OrgoSol™ buffer. The proprietary precipitation agent ensures >99% protein recovery, however precipitation may result in some loss of a protein's biological activity.
Informations de livraison: Available in two sizes. Micro kit for processing up to a total of 10 ml protein solution; Medi kit for processing up to a total of 30 ml protein solution, either in a single or multiple experiments.