Imprimer
ThermoSafe® PUR Insulated Shippers
Fournisseur: Sonoco Thermosafe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E126 E38HTOP E337 E186 E327 E366 E145
SCATE126EA 97.7 EUR
SCATE126 SCATE38HTOP SCATE337 SCATE186 SCATE327 SCATE366 SCATE145
ThermoSafe® PUR Insulated Shippers
Boîtes d'expédition et de transport
High perfomance polyurethane (PUR) insulated shippers with foam plug closure.
  • PUR offers greater insulating capability when compared to same size EPS container
  • Thick walls and outer corrugate provides protective shock absorption and cushioning during shipping and handling
  • Protects highly temperature-sensitive shipments
  • Economical and lightweight
Rigid, one-piece, three-layer laminate construction provides excellent durability. Proprietary process laminates insulation core to inner and outer corrugate walls for superior strength and temperature control for mild to extreme temperatures. Withstands crush compression in excess of 2000 kg. Includes a flexible polyurethane foam plug which seals the top, cushions and insulates contents, and vents dry ice for frozen applications.