Imprimer
Deep well plates, 24-well, Axygen®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P-DW10ML24C P-DW10ML24CS PDW10ML24C2BCS
AXYGP-DW10ML24CEA 209 EUR
AXYGP-DW10ML24C AXYGP-DW10ML24CS AXYGPDW10ML24C2BCS
Deep well plates, 24-well, Axygen®
Microplaques Microplaques deepwell
Deep well and assay plates are used for sample collection, in vitro growth chambers and longterm storage. Made of PP, they offer excellent chemical resistance and will withstand temperatures down to –80 °C. Ultra-flat surface ensures proper sealing with heat sealing film. Clear V-shaped well bottom.