Imprimer
Electronic crimpers and decappers
Fournisseur: PerkinElmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N9304501 N9304500 N9304502 N9304503
PERCN9304501EA 2670 EUR
PERCN9304501 PERCN9304500 PERCN9304502 PERCN9304503
Electronic crimpers and decappers
Pinces à sertir et pinces à dessertir
The electronic universal, precision control, power crimpers with adjustable settings are designed to provide multiple crimps on a single battery charge.
  • Rated 1500 charge cycles
  • Direct plug connection to board
  • PTC's integral to battery
  • Very low self-discharge
  • Crimper can operate while battery is charging
Certifications: Tested to EN62133 and UN DOT