Imprimer
Bottles, narrow neck, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F106200007 F106200008
BELAF106200007EA 202 EUR
BELAF106200007 BELAF106200008
Bottles, narrow neck, SP Bel-Art
Flacons Flacons en plastique
HDPE, with PP screw caps.
  • Recycling code molded into the base
  • Watertight
  • Good chemical resistance, good temperarue resistance
This leakproof design makes them ideal for packaging and storage of liquids and powders.