Imprimer
Bottles, wide neck, with screw caps, PULVIS
Fournisseur: Müller + Krempel
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MUEL14.024.27EA 87.7 EUR
MUEL14.024.27 MUEL14.024.26
Bottles, wide neck, with screw caps, PULVIS
Flacons Flacons en verre
Glass, amber.
  • Amber coloured to protect light-sensitive materials
  • For applications that do not involve high thermal stresses
  • Screw cap must be ordered seperately
Ideal for sampling, storing and transporting powders and pastes.