Imprimer
Vacuum jars, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DS5320-0910 DS5320-1212
NALGDS5320-0910EA 328 EUR
NALGDS5320-0910 NALGDS5320-1212
Vacuum jars, Nalgene®
Pots Pots à usage général
Polyetherimide, amber, transparent.
  • Excellent chemical resistance (not use with unsaturated halogenated hydrocarbons)
  • Good temperature resistance
  • Not autoclavable when used for vacuum service
These chemically-resistant jars have an amber cast and may be useful for demanding service.