Imprimer
Dye conjugates dUTP CF®
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40029 40029-1 40022 40022-1 40030 40030-1 40004 40005 40006 40007 40008 40002 40003 40002-T 40003-T 40004-T 40005-T 40006-T 40007-T 40008-T 40100 40100-T
40029.EA 413 EUR
40029. 40029-1 40022. 40022-1 40030. 40030-1 40004. 40005. 40006. 40007. 40008. 40002. 40003. 40002-T 40003-T 40004-T 40005-T 40006-T 40007-T 40008-T 40100. 40100-T
Dye conjugates dUTP CF®
Systèmes de liaison et de marquage des protéines
CF® dye-dUTP can be used for TUNEL assay, microarray, or to synthesize labelled DNA probes for in situ hybridisation and nucleic acid blotting applications.
  • For labelling fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of 8 CF® dye colours from blue to near infrared
  • Biotium’s CF® dyes have superior brightness, photostability, and are water-soluble
Biotium’s next-generation CF® dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to other fluorescent dyes. CF® dye dUTP conjugates are supplied in lyophilised form.
Attention: Note: For PCR applications, Taq polymerase should be used with dUTP conjugates, because dUTP inhibits archaeal polymerases such as Pfu and Vent®.