Imprimer
Dipping Pipettes
Fournisseur: NEUBERT VOLUME GLASSWAERE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0276-50-100 0276-50-025
NEUB0276-50-100EA 120 EUR
NEUB0276-50-100 NEUB0276-50-025
Dipping Pipettes
Pipettes Pipettes Pasteur
Dipping pipettes for sampling predefined volumes. The pipettes are immersed into the samples and filled via the open top. The pipette is emptied by tilting sideways. Calibrated to deliver EX.