Imprimer
Sample concentrators, Reacti Vap™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TS-18825 TS-18826
HYPETS-18825EA 1290 EUR
HYPETS-18825 HYPETS-18826
Sample concentrators, Reacti Vap™
Chauffe-blocs Concentrateurs d'échantillon
Reacti-Vap Evaporators (9 port)
These evaporators are precision machined gassing manifolds. They provide simple, efficient evaporation by allowing the simultaneous or separate delivery of non-reactive pressurised gas to samples. They easily connect to single block Reacti-Therm™ modules.
Reacti-Vap III Evaporators (27 port)
These evaporators triple the number of samples to evaporate. Nine needles attach to each of the three individually regulated chambers. They easily connect to Reacti- Therm™ III modules.