Imprimer
Tissue grinder, Potter-Elvehjem, PTFE, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SP886000-0018 SP886000-0019 SP886000-0020 SP886000-0022 SP886000-0024
SCERSP886000-0018EA 331 EUR
SCERSP886000-0018 SCERSP886000-0019 SCERSP886000-0020 SCERSP886000-0022 SCERSP886000-0024
Tissue grinder, Potter-Elvehjem, PTFE, KIMBLE®
Homogénéisateurs Broyeurs pour tissus
Glass tube with a PTFE pestle. Tissue grinders with capacity of 5 to 45 ml have radial serrations on the lower portion of the pestle to deliver the homogenate into the cylindrical portion of the tube.
  • Replacement components are available and interchangeable
Clearance between pestles and tubes is 0,004 to 0,006".