Imprimer
1000 and 1700 series Gastight® syringes for PSD/2 syringe pump
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8300-15 8300-20 8300-25 8300-30 8300-35 8300-40 8300-45 8300-50
HAMI8300-15EA 224 EUR
HAMI8300-15 HAMI8300-20 HAMI8300-25 HAMI8300-30 HAMI8300-35 HAMI8300-40 HAMI8300-45 HAMI8300-50
1000 and 1700 series Gastight® syringes for PSD/2 syringe pump
Seringues Seringues de chromatographie
Gastight® syringes for the PSD/2 syringe pump are available with a standard PTFE-tipped plunger and a long life ultra-high molecular weight polyethylene (UHMWPE) plunger tip. The stroke length of a PSD/2 syringe is 60 mm.
Syringe terminations:
TLL, PTFE® Luer Lock
This termination has a PTFE, male Luer taper with nickel plated brass locking hub for use with Kel-F needles, metal hub needles and universal connectors. Also, the TLL is used with Hamilton Diluters/Dispensers, OEM applications and manual operations. Autoclavable when disassembled, except on 25 ml and greater syringes. Repeated autoclaving will shorten syringe life.