Imprimer
Diagnostic vials, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
102000 102010 102060 102040 102020 102090 102050 102030
LSUK102000EA 114 EUR
LSUK102000 LSUK102010 LSUK102060 LSUK102040 LSUK102020 LSUK102090 LSUK102050 LSUK102030
Diagnostic vials, WHEATON®
Fioles
Manufactured from Type 1B neutral tubular glass. These vials are suitable for diagnostic and pharmaceutical applications.
  • Controlled bore ensures excellent fit with rubber stoppers
  • Caps and rubber stoppers made from pharmaceutical grade materials
  • Suitable for freeze-drying and autoclaving