TAE buffer, 10X solution, molecular biology grade, ultrapure
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10X TAE solution consisting of 400 mM Tris and 0,01 M EDTA in high purity dH₂O adjusted to pH 8,3. Solution is 0,2 μm filtered and dispensed into durable, square Nalgene® bottles.
- Contaminant testing:
- DNase (endo): Passes test
- DNase (exo): Passes test
- RNase: Passes test
Dilute to 1X with high purity dH₂O for a solution consisting of 40 mM Tris acetate, pH 8,3 and 1,0 mM EDTA.
Cdt: The 5 litre size is supplied in an upright carton with a convenient dispensing spout.