Imprimer
Buffers, TBS, 20X solution, pH 7,4, molecular biology grade, ultrapure
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
J75892.K2 J75892.AE J75892.K8
J75892.K2EA 139 EUR
J75892.K2 J75892.AE J75892.K8
Buffers, TBS, 20X solution, pH 7,4, molecular biology grade, ultrapure
Tampons Tampons d'électrophorèse Tampons de lavage
Used in Western blotting protocols.
  • Contaminant testing:
  • DNase (endo): Passes test
  • DNase (exo): Passes test
  • RNase: Passes test
  • Protease: Passes test
20X TBS (Tris Buffered Saline) solution consisting of 500 mM Tris, 60 mM KCl and 2,8 M NaCl in high purity dH₂O, pH 7,4. Solution is 0,2 μm filtered and dispensed into durable, square Nalgene® bottles. Dilute to 1X with high purity water for a solution consisting of 25 mM Tris, pH 7,4, 3,0 mM KCl and 140 mM NaCl. With pH (25 °C, 20X): 7,4 ±0,1.