Imprimer
Buckets, TruCLEAN™
Fournisseur: PERFEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
22-15R 22-15W 22-15B
129-0106EA 0 EUR
129-0106 129-0107 129-0108
Buckets, TruCLEAN™
Godets
Temperature resistant polypropylene buckets with stainless steel handles.
  • Buckets interlock when placed side by side
  • Non-porous, super smooth surface prevents microbial growth
  • Embossed graduations in English for accurate measure
  • Stainless steel handles will not rust, corrode or oxidize
  • Autoclavable, gamma ETO sterilization compatible
  • Impervious to most chemicals, solvents and detergents
Buckets are seated firmly within cart base eliminating any possibility of spillage during transport.
Ideal for any critical manufacturing environment. Compatible with TruCLEAN Pro Mopping Systems.