Imprimer
Cooling elements and frames
Fournisseur: DELTA T
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A100029DE A100005DE A100017DE A100006DE A100030DE A100018DE A100035 A100036 A100011 A100012 A100041
DLTTA100029DEEA 233 EUR
DLTTA100029DE DLTTA100005DE DLTTA100017DE DLTTA100006DE DLTTA100030DE DLTTA100018DE DLTTA100035 DLTTA100036 DLTTA100011 DLTTA100012 DLTTA100041
Cooling elements and frames
Refroidisseurs Sacs isothermes
Molecular medical elements for different temperature handling of samples ranges from −32 °C to +38 °C.
  • Made of PE and natural in colour
  • Sealing made of laminated aluminium film with a protected cap
  • A liquid crystal thermometer in the elements shows its current temperature
Used for transportation and storage of samples like blood products, homograft’s, cell cultures, stem cells, antibodies, proteins and other temperature sensitive medications.
Inside dimension: 210×110×30 mm