Imprimer
SPE plates, 96-well, HyperSep™, SCX
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60300-582 60300-588
HYPE60300-582EA 463 EUR
HYPE60300-582 HYPE60300-588
SPE plates, 96-well, HyperSep™, SCX
Microplaques Plaques d'extraction pour phase solide
Thermo Scientific™ HyperSep SCX Strong Cation Exchanger SPE 96-Well Plates and Individual Wells feature a strong cation exchange sorbent for extraction of charged basic compounds. The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.