Imprimer
In-line gasket filters, Gasketgard®
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WGPM0GGG2 WGPM0GGT2 WGPM0GGT4
ENTEWGPM0GGG2EA 0 EUR
ENTEWGPM0GGG2 ENTEWGPM0GGT2 ENTEWGPM0GGT4
In-line gasket filters, Gasketgard®
Filtres Systèmes de filtres en ligne
These are compact, all-metal filters that fit in place of standard ¹/₄", ³/₈" and ¹/₂" VCR® gaskets in gas distribution systems. They protect sensitive gas components, such as mass flow controllers, regulators and valves from particle damage in applications up to 400 °C.
  • Ideal for high-pressure and high-temperature applications
  • Manufactured in an ISO-certified facility
Filter and gasket: Sintered 316L stainless steel filter media welded to 316L stainless gasket; retainer: 316L stainless steel.
Particle retention: >95% removal of all particlesEn savoir plus