Imprimer
Phusion® High-Fidelity PCR Master Mix with GC Buffer
Marques : FINNZYMES REAGENTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F-532L F-532S
F-532LEA 836 EUR
F-532L F-532S
Phusion® High-Fidelity PCR Master Mix with GC Buffer
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
Phusion® High-Fidelity PCR Master Mix with GC Buffer is a convenient 2X mix containing Phusion® DNA polymerase, nucleotides and optimised GC buffer including MgCl₂. Only template and primers need to be added by the user.
Informations de livraison: Each kit contains Phusion® DNA Polymerase, 2X Phusion® GC Buffer and 400 µM of each dNTP. The master mix supplies 1,5 mM MgCl₂ and 200 µM dNTP in final reaction concentration. A separate tube of DMSO is provided with the product.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Application
Learn more