Imprimer
Fittings, flanged, Omni-Lok™
Fournisseur: OMNIFIT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
008NF16-YC5B 008NF16-YC6B 008NF32-YC5B 008NF32-YC5M 008NF32-YC6B 008NF32-YL5B 008NF16-YC5G
OMNI008NF16-YC5BEA 50.6 EUR
OMNI008NF16-YC5B OMNI008NF16-YC6B OMNI008NF32-YC5B OMNI008NF32-YC5M OMNI008NF32-YC6B OMNI008NF32-YL5B OMNI008NF16-YC5G
Fittings, flanged, Omni-Lok™
Raccords et connecteurs
Flanged tubing system is used for economical low pressure connection. A flanging washer is placed between the flange and the fitting nut. When screwed into a flat-bottom, the flange is compressesed against the bottom of the port to produce a seal.
  • Pressure rated up to 250 psi
  • Used for semi-rigid tubing and flat bottom ¼"-28 UNF ports