Imprimer
Bags, zip seal, ultra resistant, Minigrip®
Fournisseur: Flexico
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ITM024218 ITM024239 ITM024243 ITM024247 ITM024212 ITM024272 ITM024237 ITM024249
129-2352EA 153 EUR
129-2352 129-2354 129-2355 129-2356 129-2351 129-2358 129-3867 MINIITM024249
Bags, zip seal, ultra resistant, Minigrip®
Sachets Sachets d'échantillonnage
Clear PE bags, thickness 100 µm, food contact.
  • Ultra resistant for heavy or sharp products
  • High transparency and gloss
  • Suitable for food contact
  • Hanging and/or venting hole
  • Resealable, reusable, 100% recyclable
Made of polyethylene (LDPE). Zip seal designed for easy opening and high speed packing. The Minigrip® bag provides an excellent mechanical resistance and high temperature resistance (from –40 up to +60 °C).
Certifications: Suitable for food contact. Certificate available on request.