Imprimer
Jars, wide mouth, conical, graduated
Fournisseur: LAMAPLAST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
233 231 238C 238 235C 235 233C 231C
LAMA233EA 3.1 EUR
LAMA233 LAMA231 LAMA238C LAMA238 LAMA235C LAMA235 LAMA233C LAMA231C
Jars, wide mouth, conical, graduated
Pots Pots à usage général
PP, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Lid must be ordered separately
Wide mouth universal containers for creams, semi solid substances, crystals, powders and granulates.