Imprimer
Tubing, PTFE
Fournisseur: KNAUER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A0732 A0154 A0873
KNAUA0732EA 59.7 EUR
KNAUA0732 KNAUA0154 KNAUA0873
Tubing, PTFE
Tuyaux Transfer Tubing
This white PTFE tubing is characterised by its non stick surface and excellent dielectric properties.
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible