Imprimer
Hand held scrapers
Fournisseur: MICRONOVA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HHSWH-8 HHSWH-10
129-0608EA 40.8 EUR
129-0608 129-0609
Hand held scrapers
Scrapers
This hand held scraper tool is made with a seamless, one piece injection-mould that prevents bacterial growth. Designed for residue and build-up removal in clean and controlled environments.
  • Injection-moulded, seamless construction
  • Will not scratch metal surfaces
  • Made from FDA approved polypropylene
  • Suitable for indirect food contact
  • Handle shaped for strong grip
  • Autoclavable
The scraper is chemical-resistant and will not rust or corrode.