Imprimer
Pointes à filtre, faible adsorption, Corning® DeckWorks
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4135 4136 4137 4138 4139 4140
613-1696EA 617 EUR
613-1696 613-1697 613-1698 613-1699 613-1739 613-1750
Pointes à filtre, faible adsorption, Corning® DeckWorks
Pointes de pipettes
Ces pointes à barrière à faible adsorption possèdent une barrière en PE haute densité inerte afin d'éliminer la contamination pendant le transport d'aérosols lors des procédures de pipetage critiques. Les pointes ne piègent pas les liquides et n'inhibent pas la PCR en cas de surpipetage.
  • Les pointes sont fabriquées dans un environnement de moulage contrôlé pour assurer exactitude et précision entre tous les volumes de pointe
  • Apyrogènes
  • Sans ADN humain, sans RNase/DNase
  • Niveau d'assurance de stérilité de 10⁻³
Ce système est conçu pour réduire la production et le gaspillage des matières plastiques. Les supports à charnière sont fabriqués en plastique recyclé.
Corning DeckWorks barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures. DeckWorks tips will not trap liquids or inhibit PCR in the event of over-pipetting.
Corning DeckWorks proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
The entire DeckWorks system is comprised of 7 sizes of graduated pipette tips which are longer than standard tips to minimise the contamination issues often associated with traditional pipette tip styles. Sizes include 10, 20, 100, 200, extended length, 300, and 1000 µl tips. The pipette tips are made in a controlled molding environment to ensure accuracy and precision across all tip volumes. All tips are manufactured from polypropylene.
Informations de commande: Corning DeckWorks pipette tips are also available in bulk, hinged rack, reload, reload starter kit, and tip station configurations.