Imprimer
Hydrolysis unit, B-411/ E-416
Fournisseur: BUCHI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
037455
458-0014EA 0 EUR
458-0014
Hydrolysis unit, B-411/ E-416
Systèmes de digestion Systèmes de minéralisation
The B-411/ E-416 hydrolysis unit has been designed to carry out acidic hydrolysis of food and feed samples prior to solvent extraction.The hydrolysis procedure is required in a number of AOAC official methods to determine fat content of food samples including cheese, seafood, dressings, meat.
  • Choice of 4 or 6 place units
  • Simultaneous hydrolysis, filtration and rinsing of samples
  • Glass sample tube with frit matches the extraction units
  • Vacuum pump reduces filtration time
Informations de commande: Buchi V-700 vacuum pump, is required for operation and must be ordered separately from the main unit.
Informations de livraison: Supplied with digestion vessels, sample tubes, aspiration tubes, caps for digestion vessels, stoppers for tubes and a 3 mm wrench.