Imprimer
Wash bottles, wide neck
Fournisseur: SEMADENI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1164 1165
215-0255EA 13.1 EUR
215-0255 215-0256
Wash bottles, wide neck
Flacons Pissettes
LDPE, translucent
  • Wide neck for quick, safe filling, leakproof closure
  • Formed from a single piece, outlet tube can no longer become contaminated or leak. Jet tip can be cut to increase flow rate
  • Good chemical resistance, heat resistant up to +80 °C