Imprimer
96-well microplates, PP, certified, WebSeal™
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
548-3371EA 75.2 EUR
548-3371 548-2401
96-well microplates, PP, certified, WebSeal™
Microplates HTS Microplates
PP, non coated, non sterile, chromatography tested and certified. Manufactured from ultra low bleed high purity basis resin and fully lot tested by GC/MS for organic extractables. Designed for sensitive applications, lower sample concentrations, regulated laboratories, and/or TIC and full chromatograms.
  • Microplates are chemically and thermally resistant and will tolerate temperatures from −80 to +121 °C
  • SBS and ANSI standard footprint design for broad instrument compatibility
  • Ideal for pharmaceutical applications, sample collection and storage, combinatorial chemistry and HTS applications
  • U- and V- bottom well format for optimal sample recovery