Imprimer
Measuring cylinders, tall form, class A, borosilicate glass, with Batch Certificate, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
213900701 213900804 213901406 213901706 213902402 213903604 213904403 213905408 213906301
215-3607EA 33.9 EUR
215-3607 215-3608 215-3609 215-3610 215-3611 215-3612 215-3613 215-3614 215-3615
Measuring cylinders, tall form, class A, borosilicate glass, with Batch Certificate, DURAN®
Éprouvettes Éprouvettes graduées
DURAN®, borosilicate glass 3.3, clear, tall form, class A.
  • With batch certificate
  • Hexagonal glass base, with spout
  • Blue graduations with ring marks at major graduations
With printed retrace code (batch identification).