Imprimer
Microtubes à centrifuger, Axygen®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ST-150-SS ST-200-SS ST-050 ST-050-SS ST-050-SS-X ST-050-X ST-150 ST-150-SS-X ST-200 ST-200-SS-X ST-200-X ST-45 ST-45-S
212-0349EA 53.3 EUR
212-0349 212-0354 212-0299 212-0343 212-0344 212-0345 212-0347 212-0350 212-0352 212-0355 212-0356 AXYGST-45 AXYGST-45-S
Microtubes à centrifuger, Axygen®
Tubes Microtubes à centrifuger
Screw cap tubes made of PP feature a cap that provides a tight secure seal.
  • Certified RNase-/DNase-free, nonpyrogenic
  • Will withstand up to 20000×g
  • Screw caps to be ordered seperately
Screw cap tubes are available in a variety of colors with a conical or self-standing bottom.