Imprimer
Drum pumps, stainless steel
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
18908 18909 16923 16924 16925 16926
MSGA18908EA 422 EUR
MSGA18908 MSGA18909 MSGA16923 MSGA16924 MSGA16925 MSGA16926
Drum pumps, stainless steel
Pompe Pompes pour conteneur/transfert
Stainless steel drum pump with PTFE seals. Drum pump with outlet hose 1,2 m and discharge arc. for flammable liquids electrically conductive. Can be used in Zone 0 and 1. Explosion group II A.
  • Available with flexible output or angled exit