Imprimer
SPE columns, Supra-Poly® XC
Fournisseur: PerkinElmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N9306441 N9306405 N9306428 N9306440 N9306500 N9306501 N9306503
PERCN9306441EA 241 EUR
PERCN9306441 PERCN9306405 PERCN9306428 PERCN9306440 PERCN9306500 PERCN9306501 PERCN9306503
SPE columns, Supra-Poly® XC
Colonnes pour chromatographie
SPE Supra-Poly® extreme capacity (XC) are polymer based columns, polystyrene-divinylbenzene (PSDVB). Ideal for extracting polar and non-polar compounds when analysing drugs and drug metabolites in biological fluids.