Imprimer
Biocide, sterile, HydroPure
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SBT16HPW SBC56HP
CONTSBT16HPWEA 428 EUR
CONTSBT16HPW 141-1578
Biocide, sterile, HydroPure
Produits nettoyants Désinfectants
Contec HydroPure is a blend of 6% stabilised hydrogen peroxide and water for injection (WFI) or purified water. Contec HydroPure is BPR authorised at EU level and is efficacious against bacteria, fungi, moulds and yeasts.
  • Hydrogen peroxide breaks down to water and oxygen making it low in residue and suitable for product contact areas
  • Guaranteed endotoxin level of less than 0,25 EU/ml so ideal for use in product contact areas
  • Sterile filtered to 0,2 µm under Grade A airflow in a Grade B cleanroom into pre-irradiated containers
  • Trigger spray and 'bag in bottle' protected system prevents bottle contents from becoming contaminated during use
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
  • Triple-/double- bagged in linear tear packaging so each bag is easy to open even when wearing gloves
Certifications: Bactéricide EN 1276 ; fongicide EN 1650 ; bactéricide et fongicide EN 13697 ; sporicide EN 13704
Informations de commande: A non sterile version for lower grade rooms is also available.
Informations de livraison: Provided either double- or triple-bagged in linear tear packaging in linear tear packaging, the product is designed for ease of entry into pharmaceutical cleanrooms.
Cdt: The 1 L trigger spray is fitted with a protective system to protect the contents during use.
Attention: Use biocides safely. Always read the label and product information before use.
Not classed as corrosive so can be used safely in all areas of a cleanroom.