Imprimer
Anti-slip bench mats
Fournisseur: DYCEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NS02/14/1 NS02/19/1 NS02/MD/2 NS02/MD/1 NS02/14/8 NS02/14/2 NS02/SM/2 NS02/SM/3 NS02/14/3 NS02/19/2 NS02/19/3 NS02/19/8 NS02/LG/1 NS02/LG/2 NS02/LG/3 NS02/LG/8 NS02/MD/3 NS02/MD/8 NS02/SM/1 NS02/SM/8
DYCENS02/14/1EA 18.7 EUR
DYCENS02/14/1 DYCENS02/19/1 DYCENS02/MD/2 DYCENS02/MD/1 DYCENS02/14/8 DYCENS02/14/2 DYCENS02/SM/2 DYCENS02/SM/3 DYCENS02/14/3 DYCENS02/19/2 DYCENS02/19/3 DYCENS02/19/8 DYCENS02/LG/1 DYCENS02/LG/2 DYCENS02/LG/3 DYCENS02/LG/8 DYCENS02/MD/3 DYCENS02/MD/8 DYCENS02/SM/1 DYCENS02/SM/8
Anti-slip bench mats
Protection des surfaces Protections pour paillasse
These multi-purpose non-slip bench mats provide unbeatable grip have been proven to grip up to a 45° angle.
  • Easy to clean and anti-microbial
  • Heat resistant up to 50 °C
  • Non-toxic and latex-free