Imprimer
Tubing, FEP
Fournisseur: METROHM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6.1805.010 6.1805.030 6.1805.110 6.1805.080 6.1805.100 6.1805.020 6.1805.180 6.1805.060 6.1805.120 6.1805.070 6.1805.130 M6.1805.050 6.1829.020 6.1805.460 6.1805.090
METR6.1805.010EA 153 EUR
METR6.1805.010 METR6.1805.030 METR6.1805.110 METR6.1805.080 METR6.1805.100 METR6.1805.020 METR6.1805.180 METR6.1805.060 METR6.1805.120 METR6.1805.070 METR6.1805.130 JJBEM6.1805.050 METR6.1829.020 METR6.1805.460 METR6.1805.090
Tubing, FEP
Tuyaux Transfer Tubing
FEP, translucent.
  • With light and kink protection
  • With thread connections at both ends
Material allows much longer lengths and tighter tolerances than PTFE tubing.