Imprimer
Gas washing bottles, Drechsel pattern
Fournisseur: ROGO SAMPAIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
329100 329250 329500 329600
ROGO329100EA 205 EUR
ROGO329100 ROGO329250 ROGO329500 ROGO329600
Gas washing bottles, Drechsel pattern
Flacons Flacons-laveurs
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.